Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
229.00 SEK - 234.00 SEK
Specifications
For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 1 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | ||||
11877583
|
Fisherbrand™
VS10-10-1 |
10 | 40 μL |
234.00 SEK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
11807593
|
Fisherbrand™
VS10-1-1 |
1 Prep, 1 Marker | 650 μL |
234.00 SEK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
11857593
|
Fisherbrand™
VS10-12-1 |
12 | 35 μL |
234.00 SEK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
11807603
|
Fisherbrand™
VS10-16MC-1 |
16 | 25 μL |
229.00 SEK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||
11857603
|
Fisherbrand™
VS10-20-1 |
20 | 20 μL |
234.00 SEK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
11807613
|
Fisherbrand™
VS10-5-1 |
5 | 100 μL |
234.00 SEK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
11857613
|
Fisherbrand™
VS10-8MC-1 |
8 | 60 μL |
234.00 SEK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
11807623
|
Fisherbrand™
VS10-9-1 |
9 | 50 μL |
234.00 SEK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
Description
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Supplied as standard
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
Comb | |
1 Each | |
1 mm |