Learn More
Brady™ Color Polyester Laboratory Vial and Tube Labels
Easily differentiate lab samples with these B-494 color polyester labels, which are available in six color options. Pair with a versatile Brady™ portable label printer and print almost anywhere.
Brand: Brady™ 170920
This item is not returnable.
View return policy
Description
- Ideal for easily differentiating vials, PCR and microcentrifuge tubes, and slides
- Designed to withstand abrasions and exposure to chemicals and extreme temperatures, including immersion in liquid nitrogen and hot water baths
- Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces such as glass and polypropylene
- Available in six color options with a color bar across the top of the label; pre-sized for multiple applications, some options with matching color vial top label
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Gloss | |
Polyester | |
Labels | |
Cold temperature labelling;Laboratory labelling | |
-196°C | |
240 | |
-320°F to 158°F | |
0.003 in | |
25.40 mm (W) x 9.53 mm (H) x 9.53 mm (Dia) | |
1 in | |
0.375 in | |
9.53 mm | |
9.53 mm |
2011/65/EU, 2015/863 | |
Blue, White, Blue | |
Die-cut | |
Thermal Transfer | |
Non-sterile | |
BMP51;BMP53;M511;M510 | |
240 Sets / Box | |
25.40 mm (W) x 9.53 mm (H) x 9.53 mm (Dia) | |
-196°C to 70°C | |
0.0762 mm | |
0.375 in. | |
25.40 mm | |
0.25 in | |
0.375 in | |
M5-118-494-BL |