missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Chloride Hexahydrate, Reagent grade, Honeywell™

Product Code. 15691730 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86°C
Color Red
Quantity 250 g
Linear Formula CoCl2 · 6H2O
UN Number UN3077
Formula Weight 237.93g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 40mmHg (0°C)
Density 1.9g/cu.cm
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.