Learn More
Cobalt(Ii) Chloride Hexahydrate, Reagent grade, Honeywell™
Product Code. 15681730
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Poly Bottle
Chemical Identifiers
CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Specifications
Melting Point 86°C
Color Red
Quantity 1 kg
Linear Formula CoCl2 · 6H2O
UN Number UN3077
Formula Weight 237.93g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 40mmHg (0°C)
Density 1.9g/cu.cm
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate