missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPY2/MSAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14635-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to CNPY2.
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.Specifications
CNPY2/MSAP | |
Polyclonal | |
Unconjugated | |
CNPY2 | |
canopy 2 homolog (zebrafish), CNPY2, HP10390, MSAP, Protein canopy homolog 2, TMEM4, Transmembrane protein 4, ZSIG9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10330, 56530, 685814 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Flow Cytometry, Immunofluorescence | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QXT0, Q9Y2B0 | |
Cnpy2 | |
CNPY2,MSAP Fusion Protein Ag6214 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction