Learn More
Bochem™ Chromium-Plated Burette and Condenser Clamp
Click to view available options
Type:
Buret Clamp
Material:
Chromium-plated
Unit Size:
Each
Description
- Chromium plated
- PVC Coating
- Includes shaft
Specifications
Specifications
Material | Chromium-plated |
Includes | Shaft |
Type | Buret Clamp |
Diameter (Metric) Rod | 10 mm |