Learn More
Cefazolin sodium salt
Inhibits bacterial cell wall synthesis | CAS: 27164-46-1 | C14H16N8NaO4S3 | 479.504 g/mol
1495.00 SEK
Chemical Identifiers
CAS | 27164-46-1 |
---|---|
Molecular Formula | C14H16N8NaO4S3 |
Molecular Weight (g/mol) | 479.504 |
MDL Number | MFCD00056883 |
InChI Key | MTIAAUXSENDLGW-SLNAEPSVSA-N |
Synonym | Cefamedin |
PubChem CID | 131673922 |
IUPAC Name | (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
SMILES | [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] |
Description
Inhibits bacterial cell wall synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27164-46-1 | |
479.504 | |
MTIAAUXSENDLGW-SLNAEPSVSA-N | |
131673922 | |
[HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] |
C14H16N8NaO4S3 | |
MFCD00056883 | |
Cefamedin | |
(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
Specifications
Cefazolin Sodium Salt | |
27164-46-1 | |
White | |
C14H16N8NaO4S3 | |
3585038 | |
Cefamedin | |
[HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] | |
479.504 | |
476.48 |
Powder | |
170°C to 172°C | |
Light sensitive | |
MFCD00056883 | |
14,1917 | |
MTIAAUXSENDLGW-SLNAEPSVSA-N | |
(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium | |
131673922 | |
Cefazolin sodium salt |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 248-278-4
missing translation for 'rtecsNumber' : XI0390000
missing translation for 'tsca' : No
Recommended Storage : Keep cold