Learn More
CD90.2 Rat anti-Mouse, PE, Clone: 30-H12, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech PE-65088-100UG
This item is not returnable.
View return policy
Description
The CD90.2 antibody from Proteintech is a rat monoclonal antibody generated with cells of mouse CD90.2. This antibody recognizes mouse antigen. The CD90.2 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
CD90.2 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
30-H12 | |
PE | |
100 μg | |
21838 | |
IgG2b |