Learn More
CD44 Mouse anti-Human, CoraLite 594, Clone: 6F4H2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60224-100UL
Product Code. 16810906
  • 5106.85 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CD44. This antibody recognizes human, pig antigen. The CD44 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
CD44
Monoclonal
1280 μg/mL
Mouse
Primary
Human, Pig
Immunofluorescence
6F4H2
CoraLite 594
100 μL
960
IgG2a