missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CD300LF |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881484
|
Proteintech
13334-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871484
|
Proteintech
13334-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6, MIM 176883) or SHIP.Specifications
CD300LF | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8TDQ1 | |
146722 | |
CD300LF Fusion Protein Ag4150 | |
Primary | |
-20°C | |
CD300LF |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD300f, CD300LF, CLM 1, CLM1, CMRF35 like molecule 1, IgSF13, IREM 1, IREM1, NK inhibitory receptor, NKIR | |
CD300LF | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title