Learn More
CD3 Gamma Mouse anti-Human, CoraLite 488, Clone: 2E9G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60347-100UL
Description
The CD3 Gamma antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD3 Gamma. This antibody recognizes human antigen. The CD3 Gamma antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
CD3 Gamma | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
2E9G7 | |
CoraLite 488 | |
100 μL | |
917 | |
IgG2b |