Learn More
Catecholborane, 97%
Product Code. p-7046561
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
CAS: 274-07-7 | C6H4BO2 | 118.906 g/mol
Catecholborane is used to prepare B-alkylcatecholboranes. It finds application for the preparation of amides and macrocyclic lactams from carboxylic acids. It is used as a stereoselective reducing agent to convert beta-hydroxy ketones to syn 1,3-diols. Further, it reacts with alkyne through hydroboration to form trans vinyl borane, which is a precursor to Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Catecholborane is used to prepare B-alkylcatecholboranes. It finds application for the preparation of amides and macrocyclic lactams from carboxylic acids. It is used as a stereoselective reducing agent to convert beta-hydroxy ketones to syn 1,3-diols. Further, it reacts with alkyne through hydroboration to form trans vinyl borane, which is a precursor to Suzuki reaction.
Solubility
Miscible with diethyl ether, tetrahydrofuran, dichloromethane, chloroform, carbon tetrachloride, toluene, and benzene.
Notes
Moisture sensitive. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 274-07-7
Molecular Formula C6H4BO2
Molecular Weight (g/mol) 118.906
MDL Number MFCD00005846
InChI Key ZDQWVKDDJDIVAL-UHFFFAOYSA-N
Synonym catecholborane, 1,3,2-benzodioxaborole, benzo d 1,3,2 dioxaborole, 1,3,2$l^ 2-benzodioxaborole, cenmejuyoomffz-uhfffaoysa-n, catechol borane, catecholatoborane, benzcatechinborane, pyrocatecholborane
PubChem CID 6327445
IUPAC Name 1,3,2$l^{2}-benzodioxaborole
SMILES [B]1OC2=CC=CC=C2O1
Specifications
Melting Point 12°C
Density 1.125
Boiling Point 50°C (50 mmHg)
Flash Point 2°C (35°F)
Quantity 5 g
UN Number UN2924
Beilstein 972072
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with diethyl ether,tetrahydrofuran,dichloromethane,chloroform,carbon tetrachloride,toluene,and benzene.
Formula Weight 119.92
Percent Purity 97%
Chemical Name or Material Catecholborane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Catecholborane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,2-Benzodioxaborole