Learn More
Memmert™ Castor Frame (2-Part) for UF55
Castor frame for Memmert Ovens.
Brand: Memmert™ UF55R9
2564.77 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2-parts
- Height: 140 mm
Specifications
UF55 Oven | |
Castor Frame | |
140 mm |