Learn More
Hirschmann™ Capillary Tubes, Both Ends Open
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ Capillary Tubes, Both Ends Open can be utilized for a wide range of scientific and medical applications.
Brand: Hirschmann™ 9201375
693.40 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Both ends open
- Available in a wide range of diameters/lengths
Specifications
0.95 mm | |
Both ends open | |
Capillary Tube |
1.35 mm | |
75 mm |