Learn More
CaMKII delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15443-1-AP-20UL
 View more versions of this product
Product Code. 16814624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This antibody is specific to the C-term of delta chain, CaMKII delta. It can recognize most isoforms the gene encodes, from 50-72 kDa. It can recognize isoforms 2, 3, 4, 9, 11.
CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns
Specifications
Specifications
CaMKII delta
Polyclonal
Unconjugated
CAMK2D
CaM kinase II subunit delta, CaMK II subunit delta, CAMK2D, CAMKD, CaMKII delta
Rabbit
Antigen Affinity Chromatography
RUO
108058, 24246, 817
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13557
CAMK2D
CaMKII delta Fusion Protein Ag7323
20 μL
Primary
Mouse, Human, Rat