missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22274-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to CALML6.
CALML6 (CAGLP) belongs to the calmodulin family and Calglandulin subfamily. It contains 4 EF-hand domains.Specifications
CALML6 | |
Polyclonal | |
Unconjugated | |
CALML6 | |
CAGLP, Calglandulin like protein, CALGP, CALML6, calmodulin like 6, Calmodulin like protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
163688 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TD86 | |
CALML6 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction