Learn More
Calcium DL-glycerate dihydrate
Product Code. p-7044783
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
CAS: 67525-74-0 | C3H5O4 | 105.07 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in cold and hot water.
Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67525-74-0
Molecular Formula C3H5O4
Molecular Weight (g/mol) 105.07
MDL Number MFCD00149289,MFCD00149288,MFCD00149289
InChI Key RBNPOMFGQQGHHO-UWTATZPHSA-M
Synonym calciumdl-glycerate
PubChem CID 131883203
IUPAC Name calcium;2,3-dihydroxypropanoic acid;dihydrate
SMILES OC[C@@H](O)C([O-])=O
Specifications
Melting Point ∽140°C (decomposition)
Quantity 1 g
Beilstein 4614104
Merck Index 14,4483
Solubility Information Soluble in cold and hot water.
Formula Weight 286.26 (250.23 Anhydrous)
Chemical Name or Material Calcium DL-glycerate dihydrate