Learn More
Caffeine Melting Point Standard, Reagecon™
Product Code. 13562100 Shop All Reagecon Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Specifications
Packaging 1g
Quantity 1 g
compliance-icons
Product Identifier
  • MELTING POINT STANDARD CAFFEINE 235 to 238°C
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.