missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.00, Crison Instruments™

Product Code. 12081462
Click to view available options
Quantity:
250 mL
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 0°C
Color Undesignated
pH 7.00
Physical Form Liquid
Quantity 250 mL
Formulation Potassium and di-sodium Phosphates, also contains germicide
Odor Odorless
Traceability to NIST Yes
Solubility Information Complete soluble in water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.