Learn More
BRANDTECH™ VITLAB™ Class A PMP Volumetric Flasks with NS Stoppers
These Class A Volumetric Flasks with NS Stoppers, calibrated 'to contain', are an excellent break-resistant alternative to glass in the laboratory.
1171.80 SEK
Specifications
Calibration | Calibrated 'to contain' per DIN EN ISO 1042 |
---|---|
Closure Material | Polypropylene |
Color | Translucent |
Material | PMP |
Includes | Imprinted lot number, batch certificate |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | |||
11308553
|
BRANDTECH™ VITLAB™
67404 |
250 mL | 19/26 | 0.15 mL | N/A | ||||
Description
- Meet Class A tolerances per DIN EN ISO 1042
- Manufactured from crystal-clear polymethylpentene
- Supplied certificate includes mean test value and lot (lot and year imprinted on product)
- Thermal stress up to 121°C (autoclaving) does not cause permanent exceeding of the tolerance limits
- However, to preserve markings, cleaning at no higher than 60°C is recommended
- Available in a range of capacities and stopper sizes
Specifications
Calibrated 'to contain' per DIN EN ISO 1042 | |
Translucent | |
Imprinted lot number, batch certificate | |
Autoclavable |
Polypropylene | |
PMP | |
121°C max. | |
NS Stopper |