Learn More
Brady™ BBP™ Repositional Vinyl Film
BBP™ Repositional Vinyl Film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
Brand: Brady™ 118009
2134.91 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue making it ideal for applications like 5S and inventory
- Vinyl film can be thermal transfer printed
- Average outdoor durability is 5 years
- Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Acrylic, Repositionable, Pressure-sensitive | |
Gloss | |
Vinyl | |
Ideal for temporary, Removable and repositionable labels, like 5S and lean inventory label | |
5 Years | |
-40 to 180°F | |
0.005 in. | |
30.48 m | |
B30C-1125-581-YL |
Yellow | |
Film | |
Non-sterile | |
BBP30, BBP31, BBP33, BBP35, and BBP37 printers | |
1 Roll | |
-40°C to +82°C | |
100 ft. | |
1.12 in. |