Learn More
IKA™ Borosilicate Glass Coated Receiving Flask
1781.72 SEK - 3994.69 SEK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11895932
|
IKA™
0003743200 |
100 mL |
1781.72 SEK
Each |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
11805942
|
IKA™
0003743300 |
250 mL |
1838.46 SEK
Each |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
11845942
|
IKA™
0003743700 |
3000 mL |
3994.69 SEK
Each |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
11815942
|
IKA™
0003743400 |
500 mL |
2281.06 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
Specifications
Borosilicate Glass | |
Receiving Flask | |
1 Each |