Learn More
3M™ Blue Disposable Visitor Lab Coats, Series 4400
Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.
Brand:  3M™ 7000089703
 View more versions of this product
Product Code. 15324052
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Ideal for hygiene applications
  • 100% soft, breathable, and lightweight polypropylene
  • Zip fastener
  • Internal pocket
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lab Coat
Polypropylene
Zipper
Single-use
40 to 43 in.
CE Category 1
Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
2X-Large
Blue
Elastic
Serged
102 to 109 cm
Non-hazardous liquids and dust
Individual packaging