Learn More
BRAND™ Blaubrand™ Amber Borosilicate Glass Class A Volumetric Flask, with Certificate
Manufactured from Amber Borosilicate Glass and complies with DIN EN ISO 1042 standards. Includes a polypropylene stopper, but is interchangeable with amber glass stopper. The dark flask protects against deterioration of light-sensitive liquids. Delivered with individual conformity certificate.
67.05€ - 116.00€
Specifications
Calibration | Calibrated To Contain |
---|---|
Closure Material | Polypropylene |
Color | Amber |
Material | Borosilicate Glass |
No. per Case | 2 Piece |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15804978
|
BRAND™
937402 |
10 mL |
67.15€
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15814978
|
BRAND™
937403 |
20 mL |
67.05€
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15815552
|
BRAND™
937404 |
25 mL |
67.05€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
15824978
|
BRAND™
937405 |
50 mL |
68.20€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
15825552
|
BRAND™
937407 |
100 mL |
68.65€
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
15835552
|
BRAND™
937408 |
200 mL |
75.35€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
15834978
|
BRAND™
937410 |
500 mL |
88.65€
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15845552
|
BRAND™
937411 |
1000 mL |
116.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
Description
- Manufactured from Amber Borosilicate Glass.
- Complies with DIN EN ISO 1042 standards.
- Calibrated to contain (TC, In.)
- The dark glass protects against deterioration of light-sensitive liquids.
- Includes polypropylene stopper, but is interchangeable with amber glass stopper.
- Batch number and certificate included. Delivered with individual conformity certificate.
Specifications
Calibrated To Contain | |
Amber | |
2 Piece |
Polypropylene | |
Borosilicate Glass | |
A |