Learn More
Ansell BioClean-D™ Non-Sterile Cleanroom Overboots
Low-linting antistatic overboots offer added protection with a firm, elasticated fit. Available in regular or long length sizes.
3210.00 SEK
Specifications
Size | Universal |
---|---|
Material | Polypropylene |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Alert:
N/A
Description
- Lightweight low-linting CleanTough™ fabric: Lower contamination risks
- Elasticated opening: A secure fit, for enhanced personal protection
- Tie fastenings and slip-resistant sole: Secure fit and safe use
Specifications
Universal | |
Yes | |
Single-use | |
Ties | |
Bound Seams, Single Needle Stitching | |
Bound | |
Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical | |
(EU)2016/425, EN ISO 13034:2005 +A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2 | |
Dry Particulates, Liquid, Chemical-resistant, Chemical-resistant | |
Class 10 (ISO 4) |
Polypropylene | |
White | |
Non-sterile | |
Slip-resistant | |
White | |
White | |
ISO 9001:2015, ASTM F739-12 | |
30 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; five outer bags per lined carton (150 pieces) | |
Controlled Environments Shoe Covers |
N/A