Learn More
BHLHE22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25622-1-AP-150UL
 View more versions of this product
Product Code. 16894185
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.
Specifications
Specifications
BHLHE22
Polyclonal
Unconjugated
BHLHE22
Beta3, BHLHB5, BHLHE22, CAGL85, TNRC20
Rabbit
Antigen Affinity Chromatography
RUO
27319, 59058
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8C6A8, Q8NFJ8
BHLHE22
BHLHE22 Fusion Protein Ag20966
150 μL
Primary
Human, Mouse