missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHE22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25622-1-AP-20UL
This item is not returnable.
View return policy
Description
BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.Specifications
BHLHE22 | |
Polyclonal | |
Unconjugated | |
BHLHE22 | |
Beta3, BHLHB5, BHLHE22, CAGL85, TNRC20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27319, 59058 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C6A8, Q8NFJ8 | |
BHLHE22 | |
BHLHE22 Fusion Protein Ag20966 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction