Learn More
beta-Naphthol Violet, Thermo Scientific™
Product Code. 11465687
Click to view available options
Quantity:
10 g
50 g
250 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.
Solubility
Soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7143-21-7
Molecular Formula C16H9N3Na2O9S2
Molecular Weight (g/mol) 497.36
MDL Number MFCD00021457
InChI Key QMNPQXAVQXMXTD-UHFFFAOYSA-L
PubChem CID 44135658
IUPAC Name disodium;4-[(4-nitrophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[N+](=O)[O-].[Na+].[Na+]
Specifications
Solubility Information Soluble in water.
Formula Weight 497.37
Odor Odorless
Quantity 50 g
Chemical Name or Material beta-Naphthol Violet