Learn More
BD DrySlide™ Reagent Slides
Click to view available options
Quantity:
15 Pack
25 Pack
75 Pack
Type:
Indole Reagent
Nitrocefin Reagent
Oxidase Reagent
PYR Kit
Unit Size:
Pack of 15
Pack of 25
Pack of 75
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
Specifications
Specifications
Type | PYR Kit |
Quantity | 15 Pack |
Certifications/Compliance | CE |
Format | Slide |
For Use With (Application) | Used for detecting hydrolysis of PYR in the rapid presumptive identification of group A streptococci and group D enterococci from pure culture |
Storage Requirements | 15° to 30°C |