missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Battery Charger for Accumax Pipette Controller

Available in either EU or UK plug variants.

352.00 SEK

Specifications

Type Battery Charger
For Use With (Application) Charging batteries
For Use With (Equipment) Accumax Pipette Controller
View More Specs

Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15860053
View Documents
Fisherbrand™
PH01-14-1
EU plug
352.00 SEK
Each
Only null left
Estimated Shipment: 04-11-2025
Add to basket
 
Description

Description

  • Aids in the pipetting process
  • For use with the Accumax Pipette Controller
Specifications

Specifications

Battery Charger
Charging batteries
Accumax Pipette Controller
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.