Learn More
Corning™ Base Head Clamp
For Use With: Holding Rod
Brand: Corning™ 440140
670.70 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Corning offers a 2 diameter support rods. It is designed to be used with the Corning Temperature probe support clamp (Cat. No. 440141).
Note: Bosshead clamps to support rod; holding rod connects to bosshead. Temperature controller slides into holding rod.
Specifications
Holding Rod | |
Clamp |