Learn More
B23/NPM1 Mouse anti-Human, CoraLite 594, Clone: 4F12A3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60096-100UL
Product Code. 16810886
  • 450.00€ / 100µL
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Explore more special offers
This item is not returnable. View return policy
Description
Description
The B23/NPM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human B23/NPM1. This antibody recognizes human, mouse, rat antigen. The B23/NPM1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
B23/NPM1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Immunofluorescence
4F12A3
CoraLite 594
100 μL
4869
IgG1