Learn More
Azlon™ Straight-sided Beaker Set
Ideal for general laboratory use - straight stable shape
66.31 SEK - 418.00 SEK
Specifications
Material | Polypropylene |
---|---|
For Use With (Application) | Ideal for General Laboratory Use |
Type | Straight Sided |
Autoclavable | Autoclavable |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10580681
|
Azlon
BDA218MES |
50 mL, 100 mL, and 250 mL | N/A | ||||||
12483551
|
Azlon
BDA218MES |
50 mL, 100 mL, and 250 mL | N/A | ||||||
12473551
|
Azlon
BDA1198 |
500 mL, 1000 mL, and 2000 mL | N/A | ||||||
10578570
|
Azlon
BDA1199 |
10 mL, 25 mL, 50 mL, 100 mL, and 250 mL | N/A | ||||||
10293430
|
Azlon
BDA1200 |
10 mL, 25 mL, 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL | N/A | ||||||
10165824
|
Azlon
BDA300P |
10 mL, 25 mL, 50 mL, 100 mL, 250 mL, 400 mL, 500 mL, 600 mL, 1000 mL, and 2000 mL | N/A | ||||||
Description
Ideal for general laboratory use - straight stable shape
- Set of popular classic Azlon beakers
- Polypropylene material - good chemical resistance
- Can withstand temperatures up to 121°C and therefore autoclavable
- Graduations are clearly printed in indelible solvent resistant ink
Specifications
Polypropylene | |
Straight Sided | |
Yes |
Ideal for General Laboratory Use | |
Autoclavable | |
ISO 7056 |