Learn More
Axygen™ Eppendorf™ Compatible Pipet Tips
Compatible with Eppendorf and Biohit pipets
Brand: Axygen TE-1004-B
1945.08 SEK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in sterile or nonsterile forms
- e-Beam irradiated to prevent product degradation
- Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
- Graduation marks at 110 μL and 500 μL, allows visual confirmation of pipettor draw volume
- Volume: 1000 μL
- Color: Blue
Order Info
- Bulk packed: 500 Tips/Pack. 10 Packs/Cs.
- Racked: 96 Tips/Rack, 12 Racks/Pack, 3 Packs/Cs.
Specifications
1000 μL | |
Blue | |
Bulk | |
Yes |
Non-sterile | |
Eppendorf | |
Eppendorf | |
110 and 500μL |