Learn More
ATP6V1H Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26683-1-AP-150UL
 View more versions of this product
Product Code. 16895075
  • 4652.90 SEK / 150µL
Description
The ATP6V1H antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ATP6V1H. This antibody recognizes human antigen. The ATP6V1H antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
ATP6V1H
Polyclonal
Unconjugated
150 μL
51606
IgG
Western Blot, Immunoprecipitation, ELISA
367 μg/mL