Learn More
ATP6V1H Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26683-1-AP-20UL
This item is not returnable.
View return policy
Description
The ATP6V1H antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ATP6V1H. This antibody recognizes human antigen. The ATP6V1H antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
ATP6V1H | |
Polyclonal | |
Unconjugated | |
20 μL | |
51606 | |
IgG |
Western Blot, Immunoprecipitation, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human |