Learn More
Karl Hecht™ Assistent™ Square Cover Glasses for Microscopy, Boxes of 50
Preserve and protect specimens on slides. Karl Hecht™ Assistent™ Square Cover Glasses for Microscopy are manufactured from a super-transparent borosilicate glass, preventing virtually any color distortion.
2.00 €
Specifications
Product Type | Coverslip |
---|---|
Material | Glass |
Thickness (Metric) | #1 (0.13 to 0.16 mm) |
Certifications | CE |
Quantity | 50/Pk. |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10451322
|
Karl Hecht™
41000109 |
18 x 18 mm |
2.00 €
Pack of 50 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10461322
|
Karl Hecht™
41000112 |
20 x 20 mm |
2.00 €
Pack of 50 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10399341
|
Karl Hecht™
41000115 |
22 x 22 mm |
2.00 €
Pack of 50 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
Description
- 0.13 - 0.16mm thickness
- Excellent flatness
- With CE marking
- Optimum resistance to chemical attack
- Refractive index finely adapted to microscopes
- High spectral transmission
- Excellent internal glass quality with very low levels of inclusions, striae, bubbles, streaks, etc.
- In plastic boxes of 50 pieces, 20 boxes in a carboard box
Specifications
Coverslip | |
#1 (0.13 to 0.16 mm) | |
50/Pk. |
Glass | |
CE |