missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APTX Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26747-1-AP-20UL
This item is not returnable.
View return policy
Description
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2; MIM 248610), and a homodimeric dihydrolipoamide dehydrogenase (E3; MIM 238331). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene (MIM 248611) encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.Specifications
APTX | |
Polyclonal | |
Unconjugated | |
APTX | |
AOA, AOA1, aprataxin, APTX, AXA1, EAOH, EOAHA, FHA HIT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54840 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z2E3 | |
APTX | |
APTX Fusion Protein Ag24945 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction