Learn More
APTX Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26747-1-AP-150UL
 View more versions of this product
Product Code. 16855115
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2; MIM 248610), and a homodimeric dihydrolipoamide dehydrogenase (E3; MIM 238331). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene (MIM 248611) encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
Specifications
Specifications
APTX
Polyclonal
Unconjugated
APTX
AOA, AOA1, aprataxin, APTX, AXA1, EAOH, EOAHA, FHA HIT
Rabbit
Antigen Affinity Chromatography
RUO
54840
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z2E3
APTX
APTX Fusion Protein Ag24945
150 μL