Learn More
Ammonium Sulfate, 99+%, Extra Pure, SLR, Fisher Chemical™
Product Code. 11483713 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.50kg
500g
5kg
Chemical Identifiers
CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O
Specifications
Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 500 g
Formula Weight 132.13g/mol
Packaging Plastic powder jar
Product Identifier
  • Ammonium sulfate