missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10488970 Shop All Fisher Chemical Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
missing translation for 'unitSize'
1kg
2.50kg
500g
5kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 1 kg
Formula Weight 132.13g/mol
Packaging Plastic powder jar
Product Identifier
  • Ammonium sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.