missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium acetate, for analysis, ExpertQ, Scharlab
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Specifications
Specifications
CAS | 631-61-8 |
Quantity | 1 kg |
Molecular Formula | C2H7NO2 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
SMILES | [NH4+].CC([O-])=O |
IUPAC Name | ammonium acetate |
Molecular Weight (g/mol) | 77.08 |
Grade | ExpertQ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction