Learn More
Bel-Art™ SP Scienceware™ Teflon FEP Coated Aluminum Scoops
Teflon FEP Coated Aluminum Scoops are FDA compliant for food contact
1532.22 SEK - 1960.00 SEK
Specifications
Product Type Scoop
Material Aluminum
Description Sampling Scoop
Handle Type Non-Slip Grip
Color Brown
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11731495
View Documents
Bel-Art™ SP Scienceware™
H36733-0000
1 L
1532.22 SEK
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
11741495
View Documents
Bel-Art™ SP Scienceware™
H36735-0000
2 L
1960.00 SEK
Each
Only null left
Estimated Shipment: 21-08-2025
Add to basket
11751495
View Documents
Bel-Art™ SP Scienceware™
H36737-0000
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Anti-stick, corrosion resistant brown coating
  • Complies with FDA regulations for food contact
  • Handle is not coated so your grip is non-slip
Specifications
Specifications
Scoop
Sampling Scoop
Brown
FDA Compliant for Food Contact
Aluminum
Non-Slip Grip
Teflon™