Learn More
Aluminum Magnesium gauze , alloy 5056, 20 mesh woven from 0.23mm (0.009 in.) dia. wire, Thermo Scientific™
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Chemical Name or Material | Aluminum Magnesium gauze, Alloy 5056 |
Name Note | 20 mesh woven from 0.23mm (0.009 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Al:Mg; 95:5 wt% |
MDL Number | MFCD00214039 |
Quantity | 75 x 75 mm |
Mesh Size | 20 Mesh |
Odor | Odorless |
Form | Wire Cloth |