Learn More
Deltalab™ Calibrated Inoculating Loops
Use both qualitative and quantitative techniques with Deltalab™ Calibrated Inoculating Loops, Double use loops feature a loop for streaking in Petri dishes at one end; a fine colony-counting tip is located at the other end.
Brand:  Deltalab™ 302753
Product Code. 15336045
  • 154.96€ / Pack of 1200
Only null left
Estimated Shipment: 22-08-2025
Description
  • Sterilized by radiation
  • Double use loops
  • 10μL
  • Ergonomic gripping from hexagonal design
  • Calibrated loops
Specifications
Specifications
Sterile
Inoculating Loop