Learn More
Al-23 Solid Rod, Round;Diameter (mm), 5
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
400 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
250mm
300mm
400mm
600mm
800mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Chemical Name or Material | Al-23 Solid Rod |
Quantity | 400 mm |
Form | Rod |