Learn More
JP Selecta™ Stainless Steel Micro Spatula w/Flat and Spoon Ends
Used for general laboratory use.JP Selecta™ Stainless Steel Micro Spatula w/Flat and Spoon Ends is made from AISI 304 stainless steel.
7.21€ - 9.00€
Specifications
Quantity | 1 Ea. |
---|---|
Product Type | Micro Spatula |
Width (Metric) | 4 mm |
Material | Stainless Steel (304) |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12077574
|
JP Selecta™
1277100 |
100 mm |
7.21€
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
12097574
|
JP Selecta™
1277210 |
210 mm |
9.00€
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
12087574
|
JP Selecta™
1277150 |
150 mm |
9.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Made from AISI 304 stainless steel
Specifications
1 Ea. | |
Micro Spatula | |
4 mm | |
Stainless Steel (304) |