Learn More
Eppendorf™ Adapter for 5/15 mL Conical Tubes and Plates
Conical tube and plate adapter for rotor S-4xUniversal-Large.
Brand: Eppendorf™ 5920757008
3700.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2 pieces
- Adapter
Specifications
Adapter | |
5 mL/15 mL conical tubes and plates | |
For Rotor S-4xUniversal-Large | |
2 |