Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand: Socorex™ 825.0050
2647.00 SEK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of polypropylene
- With precision setting mechanism
- Single-handed volume setting
Specifications
Autoclavable | |
825 | |
Acura | |
5 to 50 μL |
Polypropylene | |
1 | |
Micropipette | |
1 Each |