Learn More
ACP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15236-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
ACP2 | |
Polyclonal | |
Unconjugated | |
ACP2 | |
acid phosphatase 2, lysosomal, ACP2, LAP, Lysosomal acid phosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11432, 24162, 53 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11117, P20611, P24638 | |
ACP2 | |
ACP2 Fusion Protein Ag7356 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |